siRNA Details
| Gene Symbol: Tktl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109534.1 | NP_001103004.1 | 6 | 727 | ||
| Gene Symbol: LOC103689980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229544.2 | XP_006229606.1 | 806 | |||

Tktl1
Genomic Map