siRNA Details
| Gene Symbol: LOC103689980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229544.2 | XP_006229606.1 | 1043 | |||
| Gene Symbol: Tktl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109534.1 | NP_001103004.1 | 7 | 964 | ||

LOC103689980
Genomic Map