siRNA Details
| Gene Symbol: Smarcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106861.1 | NP_001100331.1 | 2 | 297 | ||
| XM_006243770.2 | XP_006243832.1 | 2 | 311 | |||
| XM_006243771.2 | XP_006243833.1 | 2 | 311 | |||
| XM_006243772.2 | XP_006243834.1 | 2 | 311 | |||

Smarcc1
Genomic Map