siRNA Details
| Gene Symbol: Tex28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191103.1 | NP_001178032.1 | 454 | |||
| XM_008773612.1 | XP_008771834.1 | 559 | ||||
| Gene Symbol: LOC102551117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773614.1 | XP_008771836.1 | 3 | 559 | ||

Tex28
Genomic Map