siRNA Details
| Gene Symbol: Asb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170749.1 | NP_001164220.1 | 5 | 560 | ||
| XM_008771008.1 | XP_008769230.1 | 6 | 664 | |||
| GenBank mRNA | AW919055.1 | - | 5 | 561 | ||
| FM122942.1 | - | 5 | 547 | |||

Asb14
Genomic Map