siRNA Details
Gene Symbol: Jakmip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163277.1 | NP_001156749.1 | 23 | 3647 | ||
XM_008760004.1 | XP_008758226.1 | 27 | 4742 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Jakmip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163277.1 | NP_001156749.1 | 23 | 3647 | ||
XM_008760004.1 | XP_008758226.1 | 27 | 4742 |