siRNA Details
| Gene Symbol: Lysmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748880.3 | XP_003748928.1 | 4 | 548 | ||
| XM_008759575.1 | XP_008757797.1 | 3 | 267 | |||
| XM_008759576.1 | XP_008757798.1 | 4 | 2059 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lysmd4| Gene Symbol: Lysmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748880.3 | XP_003748928.1 | 4 | 548 | ||
| XM_008759575.1 | XP_008757797.1 | 3 | 267 | |||
| XM_008759576.1 | XP_008757798.1 | 4 | 2059 | |||