siRNA Details
| Gene Symbol: Lzts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231548.2 | XP_006231610.1 | 2 | 174 | ||
| XM_008760444.1 | XP_008758666.1 | 2 | 175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lzts2| Gene Symbol: Lzts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231548.2 | XP_006231610.1 | 2 | 174 | ||
| XM_008760444.1 | XP_008758666.1 | 2 | 175 | |||