siRNA Details
| Gene Symbol: Il20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143881.1 | NP_001137353.1 | 5 | 1039 | ||
| XM_006249724.2 | XP_006249786.1 | 6 | 1054 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il20| Gene Symbol: Il20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143881.1 | NP_001137353.1 | 5 | 1039 | ||
| XM_006249724.2 | XP_006249786.1 | 6 | 1054 | |||