siRNA Details
| Gene Symbol: Fpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001073508.4 | XP_001073508.1 | 2 | 266 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fpr2| Gene Symbol: Fpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001073508.4 | XP_001073508.1 | 2 | 266 | ||
Set Membership: |
|