siRNA Details
| Gene Symbol: Fam208a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001055921.4 | XP_001055921.3 | 6 | 1939 | ||
| XM_003751541.3 | XP_003751589.1 | 6 | 1939 | |||
| XM_006252675.1 | XP_006252737.1 | 6 | 1939 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam208a| Gene Symbol: Fam208a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001055921.4 | XP_001055921.3 | 6 | 1939 | ||
| XM_003751541.3 | XP_003751589.1 | 6 | 1939 | |||
| XM_006252675.1 | XP_006252737.1 | 6 | 1939 | |||