siRNA Details
| Gene Symbol: N4bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_214648.8 | XP_214648.5 | 7 | 3149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

N4bp1| Gene Symbol: N4bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_214648.8 | XP_214648.5 | 7 | 3149 | ||