siRNA Details
| Gene Symbol: Nkain3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109540.1 | NP_001103010.1 | 5 | 1045 | ||
| XM_008763597.1 | XP_008761819.1 | 4 | 542 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkain3| Gene Symbol: Nkain3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109540.1 | NP_001103010.1 | 5 | 1045 | ||
| XM_008763597.1 | XP_008761819.1 | 4 | 542 | |||