siRNA Details
| Gene Symbol: Hyls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729924.3 | XP_002729970.1 | 4 | 779 | ||
| XM_002729925.3 | XP_002729971.1 | 4 | 1135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hyls1| Gene Symbol: Hyls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729924.3 | XP_002729970.1 | 4 | 779 | ||
| XM_002729925.3 | XP_002729971.1 | 4 | 1135 | |||