siRNA Details
| Gene Symbol: Fpr-rs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001073753.1 | XP_001073753.1 | 1 | 808 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fpr-rs6| Gene Symbol: Fpr-rs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001073753.1 | XP_001073753.1 | 1 | 808 | ||
Set Membership: |
|