siRNA Details
| Gene Symbol: Cyp2c7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759609.1 | XP_008757831.1 | 2 | 447 | ||
| GenBank mRNA | BC097939.1 | - | 2 | 308 | 7368637 | |
| BC127503.1 | - | 2 | 364 | 7371672 | ||
| FQ218330.1 | - | 2 | 325 | |||
| M31031.1 | - | 2 | 283 | |||
| Gene Symbol: LOC100361547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759635.1 | XP_008757857.1 | 2 | 181 | ||

Cyp2c7
Genomic Map