siRNA Details
| Gene Symbol: Msrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044285.2 | NP_001037750.2 | 5 | 724 | ||
| GenBank mRNA | BC094309.1 | - | 5 | 714 | 7316479 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msrb1| Gene Symbol: Msrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044285.2 | NP_001037750.2 | 5 | 724 | ||
| GenBank mRNA | BC094309.1 | - | 5 | 714 | 7316479 | |