siRNA Details
| Gene Symbol: Nell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031070.2 | NP_112332.2 | 20 | 2298 | ||
| XM_008765797.1 | XP_008764019.1 | 19 | 2648 | |||
| XM_008765798.1 | XP_008764020.1 | 20 | 2269 | |||
| GenBank mRNA | AY089719.1 | - | 20 | 2298 | ||
| BC093617.1 | - | 19 | 2242 | 7312010 | ||
| EF110908.1 | - | 18 | 2091 | |||
| U48245.1 | - | 20 | 2298 | |||

Nell2
Genomic Map