siRNA Details
| Gene Symbol: LOC100360856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766796.1 | XP_008765018.1 | 163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100360856| Gene Symbol: LOC100360856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766796.1 | XP_008765018.1 | 163 | |||