siRNA Details
| Gene Symbol: LOC681871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759288.1 | XP_008757510.1 | 3 | 1104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC681871| Gene Symbol: LOC681871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759288.1 | XP_008757510.1 | 3 | 1104 | ||
Set Membership: |
|