siRNA Details
Gene Symbol: Gpr21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109554.1 | NP_001103024.1 | 2 | 600 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
