siRNA Details
| Gene Symbol: Cnpy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749650.3 | XP_003749698.1 | 2 | 853 | ||
| GenBank mRNA | AB190508.1 | - | 1 | 46 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnpy1| Gene Symbol: Cnpy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749650.3 | XP_003749698.1 | 2 | 853 | ||
| GenBank mRNA | AB190508.1 | - | 1 | 46 | ||