siRNA Details
| Gene Symbol: Gstz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109445.1 | NP_001102915.1 | 3 | 212 | ||
| XM_006240399.2 | XP_006240461.1 | 3 | 186 | |||
| GenBank mRNA | BC158833.1 | - | 3 | 154 | 7381517 | |
| FJ179410.1 | - | 2 | 89 | |||

Gstz1
Genomic Map