siRNA Details
| Gene Symbol: Smim12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276483.1 | NP_001263412.1 | 2 | 434 | ||
| XM_006238907.2 | XP_006238969.1 | 3 | 583 | |||
| GenBank mRNA | DV214253.1 | - | 2 | 426 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim12| Gene Symbol: Smim12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276483.1 | NP_001263412.1 | 2 | 434 | ||
| XM_006238907.2 | XP_006238969.1 | 3 | 583 | |||
| GenBank mRNA | DV214253.1 | - | 2 | 426 | ||