siRNA Details
| Gene Symbol: Pigf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239725.2 | XP_006239787.1 | 2 | 120 | ||
| XM_008764472.1 | XP_008762694.1 | 2 | 159 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pigf| Gene Symbol: Pigf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239725.2 | XP_006239787.1 | 2 | 120 | ||
| XM_008764472.1 | XP_008762694.1 | 2 | 159 | |||