siRNA Details
| Gene Symbol: Vipas39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101004.1 | NP_001094474.1 | 9, 10 | 697 | ||
| XM_006240400.2 | XP_006240462.1 | 10, 11 | 888 | |||
| GenBank mRNA | BC087099.1 | - | 9, 10 | 697 | 7192222 | |

Vipas39
Genomic Map