siRNA Details
| Gene Symbol: Tmem247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277885.1 | NP_001264814.1 | 2 | 452 | ||
| XM_008764470.1 | XP_008762692.1 | 2 | 447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem247| Gene Symbol: Tmem247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277885.1 | NP_001264814.1 | 2 | 452 | ||
| XM_008764470.1 | XP_008762692.1 | 2 | 447 | |||