siRNA Details
| Gene Symbol: LOC685273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001063116.5 | XP_001063116.2 | 1 | 695 | ||
| Gene Symbol: Snrpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271040.1 | NP_001257969.1 | 7 | 1018 | ||
| XM_006256220.2 | XP_006256282.1 | 8 | 1077 | |||

LOC685273
Genomic Map