siRNA Details
| Gene Symbol: Tm2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238417.2 | XP_006238479.1 | 1, 2 | 121 | ||
| XM_006238418.2 | XP_006238480.1 | 1, 2 | 121 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm2d1| Gene Symbol: Tm2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238417.2 | XP_006238479.1 | 1, 2 | 121 | ||
| XM_006238418.2 | XP_006238480.1 | 1, 2 | 121 | |||