siRNA Details
| Gene Symbol: Chml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109524.1 | NP_001102994.1 | 1 | 716 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chml| Gene Symbol: Chml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109524.1 | NP_001102994.1 | 1 | 716 | ||