siRNA Details
Gene Symbol: LOC100910452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773783.1 | XP_008772005.1 | 277 |
Gene Symbol: Frrs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_345535.8 | XP_345536.6 | 4 | 655 |
