siRNA Details
| Gene Symbol: L1td1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001063462.4 | XP_001063462.2 | 4 | 2627 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

L1td1| Gene Symbol: L1td1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001063462.4 | XP_001063462.2 | 4 | 2627 | ||