siRNA Details
| Gene Symbol: Reep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048047.1 | NP_001041512.1 | 3 | 274 | ||
| GenBank mRNA | BC105630.1 | - | 3 | 274 | 7320418 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reep2| Gene Symbol: Reep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048047.1 | NP_001041512.1 | 3 | 274 | ||
| GenBank mRNA | BC105630.1 | - | 3 | 274 | 7320418 | |