siRNA Details
Gene Symbol: LOC680520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001057542.4 | XP_001057542.2 | 3 | 1260 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC680520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001057542.4 | XP_001057542.2 | 3 | 1260 |