siRNA Details
| Gene Symbol: Tmed11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001071744.5 | XP_001071744.1 | 4 | 515 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmed11| Gene Symbol: Tmed11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001071744.5 | XP_001071744.1 | 4 | 515 | ||