siRNA Details
| Gene Symbol: AP2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014203.2 | NP_055018.2 | 17 | 2437 | ||
| NM_130787.2 | NP_570603.2 | 16 | 2371 | |||
| XM_011526556.2 | XP_011524858.1 | 17 | 3047 | |||
| XM_011526557.2 | XP_011524859.1 | 16 | 2976 | |||
| GenBank mRNA | AK090661.1 | - | 2 | 1357 | ||
| AK094317.1 | - | 7 | 873 | |||
| AK302005.1 | - | 13 | 1685 | |||
| AL136925.1 | - | 16 | 2237 | |||
| BC014214.2 | - | 16 | 2269 | 4111165 | ||

AP2A1
Genomic Map