siRNA Details
| Gene Symbol: Fam169b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229362.2 | XP_006229424.1 | 5 | 488 | ||
| XM_006229363.2 | XP_006229425.1 | 6 | 665 | |||
| XM_008759577.1 | XP_008757799.1 | 6 | 581 | |||
| XM_008759578.1 | XP_008757800.1 | 6 | 614 | |||
| XM_008759579.1 | XP_008757801.1 | 6 | 577 | |||

Fam169b
Genomic Map