siRNA Details
Gene Symbol: Fam169b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229362.2 | XP_006229424.1 | 5 | 427 | ||
XM_006229363.2 | XP_006229425.1 | 6 | 604 | |||
XM_008759577.1 | XP_008757799.1 | 6 | 520 | |||
XM_008759578.1 | XP_008757800.1 | 6 | 553 | |||
XM_008759579.1 | XP_008757801.1 | 6 | 516 |
