siRNA Details
| Gene Symbol: Mtmr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191096.1 | NP_001178025.1 | 7 | 799 | ||
| XM_008761329.1 | XP_008759551.1 | 7 | 644 | |||
| XM_008761330.1 | XP_008759552.1 | 7 | 1661 | |||
| GenBank mRNA | BC085792.1 | - | 7 | 773 | ||

Mtmr11
Genomic Map