siRNA Details
Gene Symbol: Mtmr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191096.1 | NP_001178025.1 | 14 | 1496 | ||
XM_008761329.1 | XP_008759551.1 | 14 | 1341 | |||
XM_008761330.1 | XP_008759552.1 | 14 | 2358 | |||
GenBank mRNA | BC085792.1 | - | 13 | 1381 |
