siRNA Details
Gene Symbol: Rad21l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001072159.4 | XP_001072159.3 | 6 | 718 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
