siRNA Details
| Gene Symbol: FEZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042548.1 | NP_001036013.1 | 5 | 912 | ||
| NM_005102.2 | NP_005093.2 | 5 | 912 | |||
| XM_006712153.2 | XP_006712216.1 | 5 | 734 | |||
| GenBank mRNA | AB076184.1 | - | 5 | 904 | ||
| AB209719.1 | - | 5 | 711 | |||
| AF113124.1 | - | 5 | 811 | |||
| AK025274.1 | - | 4 | 592 | |||
| BC037566.1 | - | 5 | 877 | |||
| BC047020.1 | - | 5 | 863 | |||
| BC089390.1 | - | 5 | 858 | |||
| BC110386.1 | - | 5 | 867 | 5183735 | ||
| BX537785.1 | - | 6 | 838 | |||
| U60061.1 | - | 2 | 265 | |||
| U69140.1 | - | 4 | 553 | |||

FEZ2
Genomic Map