siRNA Details
| Gene Symbol: Txlna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127633.1 | NP_001121105.1 | 5, 6 | 880 | ||
| XM_006238972.2 | XP_006239034.1 | 5, 6 | 887 | |||
| GenBank mRNA | BC166486.1 | - | 5, 6 | 880 | 7099983 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txlna| Gene Symbol: Txlna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127633.1 | NP_001121105.1 | 5, 6 | 880 | ||
| XM_006238972.2 | XP_006239034.1 | 5, 6 | 887 | |||
| GenBank mRNA | BC166486.1 | - | 5, 6 | 880 | 7099983 | |