siRNA Details
| Gene Symbol: FEZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042548.1 | NP_001036013.1 | 2 | 327 | ||
| NM_005102.2 | NP_005093.2 | 2 | 327 | |||
| XM_006712153.2 | XP_006712216.1 | 2 | 149 | |||
| GenBank mRNA | AB076184.1 | - | 2 | 319 | ||
| AB209719.1 | - | 2 | 126 | |||
| AF113124.1 | - | 2 | 226 | |||
| BC018032.1 | - | 2 | 306 | |||
| BC037566.1 | - | 2 | 292 | |||
| BC047020.1 | - | 2 | 278 | |||
| BC054043.1 | - | 2 | 261 | |||
| BC064484.1 | - | 2 | 241 | |||
| BC089390.1 | - | 2 | 273 | |||
| BC110386.1 | - | 2 | 282 | 5183735 | ||
| BX537785.1 | - | 2 | 167 | |||
| L17328.1 | - | 1 | 555 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_199448, NM_001285940, NM_001285949, NM_001285946, AK157221, and others.

FEZ2
Genomic Map