siRNA Details
| Gene Symbol: LOC690806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135576.2 | NP_001129048.1 | 864 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690806| Gene Symbol: LOC690806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135576.2 | NP_001129048.1 | 864 | |||