siRNA Details
| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126299.2 | NP_001119771.2 | 9 | 2868 | ||
| XM_006254412.2 | XP_006254474.1 | 11 | 3481 | |||
| XM_006254413.2 | XP_006254475.1 | 11 | 3291 | |||
| XM_006254414.2 | XP_006254476.1 | 10 | 3290 | |||
| GenBank mRNA | BC166441.1 | - | 9 | 2868 | 7461161 | |

Esco1
Genomic Map