siRNA Details
| Gene Symbol: Clnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251106.2 | XP_006251168.1 | 8 | 708 | ||
| XM_006251108.2 | XP_006251170.1 | 8 | 708 | |||
| XM_008770228.1 | XP_008768450.1 | 5 | 554 | |||
| XM_008770229.1 | XP_008768451.1 | 5 | 554 | |||
| XM_008770230.1 | XP_008768452.1 | 5 | 554 | |||

Clnk
Genomic Map