siRNA Details
| Gene Symbol: LOC103689981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758849.1 | XP_008757071.1 | 347 | |||
| Gene Symbol: LOC100910798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750426.3 | XP_003750474.2 | 347 | |||
| Gene Symbol: Vom2r19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099492.1 | NP_001092962.1 | 2 | 347 | ||
More Information
Set Membership: |
|

LOC103689981
Genomic Map