siRNA Details
| Gene Symbol: Ppfia4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080409.1 | NP_536334.1 | 2 | 184 | ||
| XM_008769527.1 | XP_008767749.1 | 2 | 446 | |||
| XM_008769528.1 | XP_008767750.1 | 2 | 446 | |||
| XM_008769529.1 | XP_008767751.1 | 2 | 446 | |||
| GenBank mRNA | BF285985.1 | - | 2 | 185 | ||

Ppfia4
Genomic Map