siRNA Details
| Gene Symbol: Dazl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109414.1 | NP_001102884.1 | 6 | 640 | ||
| XM_006244477.2 | XP_006244539.1 | 6 | 493 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dazl| Gene Symbol: Dazl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109414.1 | NP_001102884.1 | 6 | 640 | ||
| XM_006244477.2 | XP_006244539.1 | 6 | 493 | |||